Methyl viologen dichloride hydrate 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Methyl Viologen hydrate, Gramoxone, Paraquat dichloride hydrate, 1,1′-Dimethyl-4,4′-bipyridinium dichloride
227320010
227320050
ACRO227320010EA
75.7
EUR
ACRO227320010
ACRO227320050
Methyl viologen dichloride hydrate 98%
Methyl viologen dichloride hydrate
CAS No.: 1910-42-5
Formel:
C₁₂H₁₄Cl₂N₂·nH₂O Schmelzpunkt: >300 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00150001 CAS-Nummer: 1910-42-5 UN-Nummer: 2811 ADR: 6.1,I |
Spezifikation Testergebnisse
Appearance | Beige to pink to brown Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % (on dry substance) |
Water | ≤18 % (K.F.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...