N-Acetyl-D-galactosamine 97%
Lieferant: Thermo Fisher Scientific
225850010
225855000
ACRO225850010EA
155
EUR
ACRO225850010
ACRO225855000
N-Acetyl-D-galactosamine 97%
N-Acetyl-D-galactosamine
CAS No.: 14215-68-0
Appearance: White to off-white Crystalline powder
Formel:
C₈H₁₅NO₆ Schmelzpunkt: 158…162 °C Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00065372 CAS-Nummer: 14215-68-0 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤7.0 % (K.F.) |
Solubility | (10 % in water) Clear colorless |
HPLC | ≥96.0 % (ELSD) |
Specific optical rotation | +80° to +90° (20°C, 589 nm) (c=1, water) (after 5 h, on dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...