Polyethylenglycol 1.450 NF
Lieferant: Spectrum Chemical
Synonyme:
PEG 1,450, Ethylene glycol polymer 1450, PEG 1450
P1492-12KGBL
P1492-2.5KG
P1492-45KGBL
SPCMP1492-12KGBLEA
1110
EUR
SPCMP1492-12KGBL
SPCMP1492-2.5KG
SPCMP1492-45KGBL
Polyethylenglycol 1.450 NF
Polyethylenglycol 1.450
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formel:
H(OCH₂CH₂)nOH Molekulargewicht: 1450 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00081839 CAS-Nummer: 25322-68-3 EINECS: 500-038-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...