4-(Cyanomethyl)phenylboronic acid 98%

Lieferant: Thermo Fisher Scientific
Achtung

L19955.03 L19955.06
L19955.03EA 103 EUR
L19955.03 L19955.06
4-(Cyanomethyl)phenylboronic acid 98%
4-(Cyanomethyl)phenylboronic acid

Purity: 98% CAS No.: 91983-26-5 Molecular Formula: C8H8BNO2 Formula Weight: 160.97 Melting Point: ca 196-200° Beilstein Reference: 3031323 MDL No.: MFCD01632200

Formel: C₈H₈BNO₂
Schmelzpunkt: ∼ 196…200 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01632200
CAS-Nummer: 91983-26-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR