4-Ethynyl-benzoic acid sodium salt 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Sodium 4-ethynylbenzoate

L19292.03 L19292.06
L19292.03EA 99.1 EUR
L19292.03 L19292.06
4-Ethynyl-benzoic acid sodium salt 97%
4-Ethynyl-benzoic acid sodium salt

Purity: 97% CAS No.: 144693-65-2 Molecular Formula: C9H5NaO2 Formula Weight: 168.13 Beilstein Reference: 5836538 MDL No.: MFCD03426026

Formel: C₉H₅NaO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03426026
CAS-Nummer: 144693-65-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR