Potassium trans-β-styryltrifluoroborate 98%

Lieferant: Thermo Fisher Scientific
Achtung

L17971.03 L17971.06
L17971.03EA 79.6 EUR
L17971.03 L17971.06
Potassium trans-β-styryltrifluoroborate 98%
Potassium trans-beta-styryltrifluoroborate

Purity: 98% CAS No.: 201852-49-5 Molecular Formula: C8H7BF3K Formula Weight: 210.05 Melting Point: 300° Beilstein Reference: 7782992 MDL No.: MFCD02093981

Formel: C₈H₇BF₃K
Schmelzpunkt: >300 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02093981
CAS-Nummer: 201852-49-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR