Cyclohexyl-p-toluolsulfonat 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Cyclohexyl 4-methylbenzenesulphonate, Cyclohexyl Tosylate, p-Toluenesulphonic acid Cyclohexyl Ester
L17952.06
L17952.14
L17952.06EA
50
EUR
L17952.06
L17952.14
Cyclohexyl-p-toluolsulfonat 97%
Cyclohexyl-p-toluenesulphonate
Purity: 97%
CAS No.: 953-91-3
Molecular Formula: C13H18O3S
Formula Weight: 254.35
Melting Point: 45-46°
Beilstein Reference: 2217391
MDL No.: MFCD00014291
Formel:
C₁₃H₁₈O₃S Schmelzpunkt: 45…46 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00014291 CAS-Nummer: 953-91-3 EINECS: 213-468-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...