Ishikawa′s Reagenz ≥90%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-(Diethylamino)-1,1,2,3,3,3-hexafluoropropane, Hexafluoropropene diethylamine, N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine

L16738.14 L16738.22
L16738.14EA 79.8 EUR
L16738.14 L16738.22
Ishikawa′s Reagenz ≥90%
Ishikawa's reagent

Purity: 90+% CAS No.: 309-88-6 Molecular Formula: C7H11F6N Formula Weight: 223.16 Flash Point: 14°(57°F) Refractive Index: 1.3500 Boiling Point: 56-57°/58mm UN No.: UN2924 MDL No.: MFCD00054683

Formel: C₇H₁₁F₆N
Siedepunkt: 56…57 °C (58 mmHg)
Dichte: 1.230
Flammpunkt: 14 °C (57 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00060264
CAS-Nummer: 309-88-6
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR