S-Tritylcystein 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: H-Cys(Trt)-OH

L14384.03 L14384.06
L14384.03EA 28 EUR
L14384.03 L14384.06
S-Tritylcystein 97%
S-Trityl-L-cysteine

Purity: 97% CAS No.: 2799-07-7 Molecular Formula: C22H21NO2S Formula Weight: 363.48 Melting Point: ca 185° dec. Beilstein Reference: 2339626 Optical Rotation: +84° (c=1 in acetic acid) MDL No.: MFCD00002611

Formel: C₂₂H₂₁NO₂S
Schmelzpunkt: ∼ 185 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00002611
CAS-Nummer: 2799-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR