1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
1-O-Acetyl-2,3,5-Tri-O-benzoyl-beta-D-Ribofuranose, (2S,3R,4R,5R)-2-Acetoxy-5-((benzoyloxy)methyl)tetrahydrofuran-3,4-diyl dibenzoate, β-D-Ribofuranose 1-Acetate 2,3,5-Tribenzoate, 2,3,5-Tri-o-Benzoyl-1-o-Acetyl-β-D-Ribofuranose, 1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose, β-D-Ribofuranose 1-acetate 2,3,5-tribenzoate
Purity: 98%
CAS No.: 6974-32-9
Molecular Formula: C28H24O9
Formula Weight: 504.50
Melting Point: 128-130°
Beilstein Reference: 100243
Optical Rotation: +44° (c=0.5 in chloroform)
MDL No.: MFCD00005357
Formel:
C₂₈H₂₄O₉ Molekulargewicht: 504.5 g/mol Schmelzpunkt: 128…130 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00005357 CAS-Nummer: 6974-32-9 EINECS: 230-220-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...