1-Pyrrolidinecarbonyl chloride 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97%
CAS No.: 1192-63-8
Molecular Formula: C5H8ClNO
Formula Weight: 133.58
Storage Temperature: 2°C to 8°C
Flash Point: 97°(206°F)
Refractive Index: 1.4940
Boiling Point: 75°/1mm
Beilstein Reference: 110281
UN No.: UN3265
MDL No.: MFCD00051321
Formel:
C₅H₈ClNO Siedepunkt: 75 °C (1 mmHg) Dichte: 1,209 Flammpunkt: 97 °C (206 °F) |
MDL Number:
MFCD00051321 CAS-Nummer: 1192-63-8 EINECS: 214-758-7 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...