2,2',5,5'-Tetramethylbiphenyl 98%

Lieferant: Thermo Fisher Scientific

L13330.03 L13330.06
L13330.03EA 70.8 EUR
L13330.03 L13330.06
2,2',5,5'-Tetramethylbiphenyl 98%
2,2',5,5'-Tetramethylbiphenyl

Purity: 98% CAS No.: 3075-84-1 Molecular Formula: C16H18 Formula Weight: 210.32 Melting Point: 48-51° Boiling Point: 284°/732mm Beilstein Reference: 2046200 MDL No.: MFCD00151846

Formel: C₁₆H₁₈
Siedepunkt: 284 °C (732 mmHg)
Schmelzpunkt: 48…51 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00151846
CAS-Nummer: 3075-84-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR