1-Phenyl-1,4-pentanedione 96%
Lieferant: Thermo Fisher Scientific
L13151.06
L13151.03
L13151.06EA
135
EUR
L13151.06
L13151.03
1-Phenyl-1,4-pentanedione 96%
1-Phenyl-1,4-pentanedione
Purity: 96%
CAS No.: 583-05-1
Molecular Formula: C11H12O2
Formula Weight: 176.22
Refractive Index: 1.5330
Boiling Point: 158-162°/12mm
Beilstein Reference: 908230
MDL No.: MFCD00092889
Formel:
C₁₁H₁₂O₂ Siedepunkt: 158…162 °C (12 mmHg) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00092889 CAS-Nummer: 583-05-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...