L-Leucine ethyl ester hydrochloride 97%

Lieferant: Thermo Fisher Scientific

Synonyme: H-Leu-OEt.HCl

L13147.06 L13147.14
L13147.06EA 31.3 EUR
L13147.06 L13147.14
L-Leucine ethyl ester hydrochloride 97%
L-Leucine ethyl ester hydrochloride

Purity: 97% CAS No.: 2743-40-0 Molecular Formula: C8H17NO2.HCl Formula Weight: 195.69 Odour: Odourless Melting Point: 134-137° Beilstein Reference: 3994312 Optical Rotation: +18° (c=5 in ethanol) MDL No.: MFCD00034879

Formel: C₈H₁₇NO₂
Molekulargewicht: 195.69 g/mol
Schmelzpunkt: 134…137 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00034879
CAS-Nummer: 2743-40-0
EINECS: 220-375-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR