Ethyl-2-nitropropionate 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L12261.03
L12261.06
L12261.03EA
40.8
EUR
L12261.03
L12261.06
Ethyl-2-nitropropionate 96%
Ethyl-2-nitropropionate
Purity: 96%
CAS No.: 2531-80-8
Molecular Formula: C5H9NO4
Formula Weight: 147.13
Flash Point: 89°(192°F)
Refractive Index: 1.4210
Boiling Point: 190-195°
Beilstein Reference: 774734
MDL No.: MFCD00024818
Formel:
C₅H₉NO₄ Siedepunkt: 190…195 °C Dichte: 1.130 Flammpunkt: 89 °C (192 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00024818 CAS-Nummer: 2531-80-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...