3-Trifluoromethylbutyric acid 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L12160.03
L12160.06
L12160.03EA
56
EUR
L12160.03
L12160.06
3-Trifluoromethylbutyric acid 97%
3-Trifluoromethylbutyric acid
Purity: 97%
CAS No.: 348-75-4
Molecular Formula: C5H7F3O2
Formula Weight: 156.11
Refractive Index: 1.3590
Odour: Pungent
Boiling Point: 90-91°/25mm
Beilstein Reference: 1760574
UN No.: UN3265
MDL No.: MFCD00039533
Formel:
C₅H₇F₃O₂ Siedepunkt: 90…91 °C (25 mmHg) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00039533 CAS-Nummer: 348-75-4 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...