n-Propyl bromoacetate 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L11951.06
L11951.14
L11951.06EA
36.7
EUR
L11951.06
L11951.14
n-Propyl bromoacetate 97%
n-Propyl bromoacetate
Purity: 97%
CAS No.: 35223-80-4
Molecular Formula: BrCH2CO2(CH2)2CH3
Formula Weight: 181.04
Flash Point: 82°(179°F)
Refractive Index: 1.4510
Boiling Point: 175-176°
Beilstein Reference: 1745564
UN No.: UN2922
MDL No.: MFCD00075247
Formel:
BrCH₂CO₂(CH₂)₂CH₃ Siedepunkt: 175…176 °C Dichte: 1.399 Flammpunkt: 82 °C (179 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00075247 CAS-Nummer: 35223-80-4 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...