N-Acetyl-L-alanine 96%
Lieferant: Thermo Fisher Scientific
Synonyme:
Ac-Ala-OH, Acetyl-L-alanine
L11811.03
L11811.06
L11811.03EA
36.4
EUR
L11811.03
L11811.06
N-Acetyl-L-alanine 96%
N-Acetyl-L-alanine
Purity: 96%
CAS No.: 97-69-8
Molecular Formula: C5H9NO3
Formula Weight: 131.13
Storage Temperature: 2°C to 8°C
Melting Point: 122-128°
Beilstein Reference: 1722932
Optical Rotation: -64° (c=2 in water)
MDL No.: MFCD00063132
Formel:
C₅H₉NO₃ Schmelzpunkt: 122…128 °C |
MDL Number:
MFCD00063132 CAS-Nummer: 97-69-8 EINECS: 202-602-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...