1-Thio-β-D-glucose tetraacetate ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
(2R,3R,4S,5R,6S)-2-(Acetoxymethyl)-6-mercaptotetrahydro-2H-pyran-3,4,5-triyl triacetate
L11404.03
L11404.MD
L11404.03EA
121
EUR
L11404.03
L11404.MD
1-Thio-β-D-glucose tetraacetate ≥98%
1-Thio-beta-D-glucose tetraacetate
Purity: 99%
CAS No.: 19879-84-6
Molecular Formula: C14H20O9S
Formula Weight: 364.37
Storage Temperature: 2°C to 8°C
Melting Point: 115-117°
Beilstein Reference: 1440689
Optical Rotation: +5.5° ± 1° (c=2, chloroform)
MDL No.: MFCD00063271
Formel:
C₁₄H₂₀O₉S Schmelzpunkt: 115…117 °C |
MDL Number:
MFCD00063271 CAS-Nummer: 19879-84-6 EINECS: 243-392-0 |