N-Acetyl-DL-alanine ≥95%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Acetyl-DL-2-aminopropionic Acid, Ac-DL-Ala-OH, N-Ac-DL-Ala-OH, N-Acetyl-DL-2-aminopropionic acid, N-Acetyl-DL-Alanine, 2-Acetylamino-propionic acid

L10329.06 L10329.14
L10329.06EA 23.1 EUR
L10329.06 L10329.14
N-Acetyl-DL-alanine ≥95%
N-Acetyl-DL-alanine

Purity: 97+% CAS No.: 1115-69-1 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Storage Temperature: 2°C to 8°C Melting Point: 135-139° Beilstein Reference: 774333 MDL No.: MFCD00037238

Formel: C₅H₉NO₃
Molekulargewicht: 131.13 g/mol
Schmelzpunkt: 135…139 °C
MDL Number: MFCD00037238
CAS-Nummer: 1115-69-1
EINECS: 214-229-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR