Methyl-2-acetamidoacrylat 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Acetamidoacrylic Acid Methyl Ester, Methyl 2-acetamidoacrylate
L10049.03
L10049.06
L10049.03EA
76.4
EUR
L10049.03
L10049.06
Methyl-2-acetamidoacrylat 97%
Methyl-2-acetamidoacrylate
Purity: 99%
CAS No.: 35356-70-8
Molecular Formula: C6H9NO3
Formula Weight: 143.14
Storage Temperature: 2°C to 8°C
Melting Point: 49-51°
Boiling Point: 103-104°/8mm
Beilstein Reference: 1812208
MDL No.: MFCD00013394
Formel:
C₆H₉NO₃ Siedepunkt: 103…104 °C (8 mmHg) Schmelzpunkt: 49…51 °C |
MDL Number:
MFCD00013394 CAS-Nummer: 35356-70-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...