1,1,1-Trifluoro-2,3-epoxypropane 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98%
CAS No.: 359-41-1
Molecular Formula: C3H3F3O
Formula Weight: 112.05
Storage Temperature: 2°C to 8°C
Flash Point: -40°(-40°F)
Refractive Index: 1.297
Boiling Point: 38-40°
Beilstein Reference: 103730
UN No.: UN1992
MDL No.: MFCD00041506
Formel:
C₃H₃F₃O Siedepunkt: 38…40 °C Dichte: 1,300 Flammpunkt: –40 °C (–40 °F) |
MDL Number:
MFCD00041506 CAS-Nummer: 359-41-1 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...