Fast blue BB salt
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Benzoylamino-2,5-diethoxybenzenediazonium chloride hemi(zinc chloride) salt, 4-Amino-2,5-diethoxybenzanilide diazotated zinc double salt, Fast Blue BB Salt hemi(zinc chloride) salt
CAS No.: 5486-84-0
Molecular Formula: C34H36Cl4N6O6Zn
Formula Weight: 831.89
Storage Temperature: -30°C to -10°C
Odour: Odourless
Melting Point: ca 157° dec.
Beilstein Reference: 1845288
MDL No.: MFCD00074765
Formel:
C₃₄H₃₆Cl₄N₆O₆Zn Molekulargewicht: 831.89 g/mol Schmelzpunkt: ∼ 157 °C (Zers.) |
MDL Number:
MFCD00074765 CAS-Nummer: 5486-84-0 EINECS: 226-817-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...