N-(tert-Butoxycarbonyl)-D-leucine hydrate ≥98%
Lieferant: Thermo Fisher Scientific
L09124.03
L09124.06
L09124.03EA
24.9
EUR
L09124.03
L09124.06
N-(tert-Butoxycarbonyl)-D-leucine hydrate ≥98%
N-(tert-Butoxycarbonyl)-D-leucine hydrate
Purity: 98+%
CAS No.: 16937-99-8
Molecular Formula: C11H21NO4.xH2O
Formula Weight: 231.29(anhy)
Melting Point: 80-84°
Beilstein Reference: 2331060
Optical Rotation: +25° (c=2 in acetic acid)
MDL No.: MFCD00065583
Formel:
C₁₁H₂₁NO₄ Schmelzpunkt: 80…84 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00038294 CAS-Nummer: 16937-99-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...