(S)-(+)-S-Methyl-S-phenylsulphoximine 97%
Lieferant: Thermo Fisher Scientific
L08618.03
L08618.MD
L08618.03EA
227
EUR
L08618.03
L08618.MD
(S)-(+)-S-Methyl-S-phenylsulphoximine 97%
(S)-(+)-S-Methyl-S-phenylsulphoximine
Purity: 97%
CAS No.: 33903-50-3
Molecular Formula: C7H9NOS
Formula Weight: 155.22
Melting Point: 30-32°
Beilstein Reference: 5245633
Optical Rotation: +36° (c=1 in acetone)
MDL No.: MFCD00151461
Formel:
C₇H₉NOS Schmelzpunkt: 30…32 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00151461 CAS-Nummer: 33903-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...