Aleuritic acid 95%
Lieferant: Thermo Fisher Scientific
L08542.06
L08542.14
L08542.06EA
57.6
EUR
L08542.06
L08542.14
Aleuritic acid 95%
Aleuritic acid
Purity: 95%
CAS No.: 533-87-9
Molecular Formula: C16H32O5
Formula Weight: 304.43
Odour: Odourless
Melting Point: 97-101°
Beilstein Reference: 1727724
MDL No.: MFCD00135596
Formel:
C₁₆H₃₂O₅ Schmelzpunkt: 97…101 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00135596 CAS-Nummer: 533-87-9 EINECS: 208-578-8 Merck Index: 14,231 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...