1-Methyl-3-n-propyl-2-pyrazolin-5-one 97%

Lieferant: Thermo Fisher Scientific

L05425.03 L05425.06
L05425.03EA 27.6 EUR
L05425.03 L05425.06
1-Methyl-3-n-propyl-2-pyrazolin-5-one 97%
1-Methyl-3-n-propyl-2-pyrazolin-5-one

Purity: 97% CAS No.: 31272-04-5 Molecular Formula: C7H12N2O Formula Weight: 140.19 Melting Point: 114-118° Beilstein Reference: 637013 MDL No.: MFCD00051849

Formel: C₇H₁₂N₂O
Schmelzpunkt: 114…118 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00051849
CAS-Nummer: 31272-04-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR