N,N-Bis(2-Chlorethyl)-p-Toluolsulfonamid 90%, tech.
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N,N-Bis(2-chloroethyl)-4-methylbenzenesulphonamide
L05296.06
L05296.14
L05296.06EA
25.4
EUR
L05296.06
L05296.14
N,N-Bis(2-Chlorethyl)-p-Toluolsulfonamid 90%, tech.
N,N-Bis(2-Chlorethyl)-p-Toluolsulfonamid
Purity: 90%
CAS No.: 42137-88-2
Molecular Formula: C11H15Cl2NO2S
Formula Weight: 296.22
Melting Point: 45-47°
Beilstein Reference: 2134258
MDL No.: MFCD00018944
Formel:
C₁₁H₁₅Cl₂NO₂S Schmelzpunkt: 45…47 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00018944 CAS-Nummer: 42137-88-2 EINECS: 255-677-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...