Dimethyl allylmalonate 97%

Lieferant: Thermo Fisher Scientific

Synonyme: Dimethyl 2-allylmalonate

L02376.09 L02376.18
L02376.09EA 49.2 EUR
L02376.09 L02376.18
Dimethyl allylmalonate 97%
Dimethyl allylmalonate

Purity: 97% CAS No.: 40637-56-7 Molecular Formula: C8H12O4 Formula Weight: 172.18 Flash Point: 95°(203°F) Refractive Index: 1.4350 Melting Point: -80° Boiling Point: 207° Beilstein Reference: 1772822 MDL No.: MFCD00014959

Formel: C₈H₁₂O₄
Siedepunkt: 207 °C
Schmelzpunkt: < –80 °C
Dichte: 1.074
Flammpunkt: 95 °C (203 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00014959
CAS-Nummer: 40637-56-7
EINECS: 255-017-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR