Chelidamic acid hydrate 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1,4-Dihydro-4-oxo-2,6-pyridinedicarboxylic acid hydrate, 4-Oxo-1,4-dihydropyridine-2,6-dicarboxylic acid hydrate
L00782.03
L00782.06
L00782.03EA
34.2
EUR
L00782.03
L00782.06
Chelidamic acid hydrate 95%
Chelidamic acid hydrate
Purity: 95%
CAS No.: 138-60-3
Molecular Formula: C7H5NO5.xH2O
Formula Weight: 183.12(anhy)
Melting Point: ca 260° dec.
MDL No.: MFCD00066478
Formel:
C₇H₅NO₅·xH₂O Schmelzpunkt: ∼ 260 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00066478 CAS-Nummer: 138-60-3 EINECS: 205-335-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...