Diethyl-benzylidenmalonat 98%

Lieferant: Thermo Fisher Scientific

Synonyme: Diethyl Benzalmalonate, Benzalmalonic Acid Diethyl Ester, Benzylidenemalonic Acid Diethyl Ester

L00561.14 L00561.22
L00561.14EA 37.3 EUR
L00561.14 L00561.22
Diethyl-benzylidenmalonat 98%
Diethyl benzylidenemalonate

Purity: 98% CAS No.: 5292-53-5 Molecular Formula: C14H16O4 Formula Weight: 248.28 Flash Point: 110°(230°F) Refractive Index: 1.5360 Melting Point: 28-31° Boiling Point: 215-217°/30mm Beilstein Reference: 528894 MDL No.: MFCD00009149

Formel: C₁₄H₁₆O₄
Siedepunkt: 215…217 °C (30 mmHg)
Schmelzpunkt: 28…31 °C
Dichte: 1.107
Flammpunkt: >110 °C (230 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00009149
CAS-Nummer: 5292-53-5
EINECS: 226-137-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR