Spectinomycin sulphate 93%

Lieferant: Thermo Fisher Scientific
Gefahr

J66034.03 J66034.06
J66034.03EA 43.9 EUR
J66034.03 J66034.06
Spectinomycin sulphate 93%
Spectinomycin sulphate

This spectinomycin sulfate antibiotic acts against Gram-negative bacteria.


  • White powder


Soluble in DMSO or water

Formel: C₁₄H₂₄N₂O₇·H₂SO₄
Siedepunkt: 583 °C
Flammpunkt: >110 °C (230 °F)
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00270185
CAS-Nummer: 23312-56-3
EINECS: 245-578-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR