Clotrimazol

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-(2-Chlorotrityl)imidazole, 1-[(2-Chlorophenyl)diphenylmethyl]imidazole

J63895.06 J63895.14
J63895.06EA 92 EUR
J63895.06 J63895.14
Clotrimazol
Clotrimazole

CAS No.: 23593-75-1 Molecular Formula: C22H17ClN2 Formula Weight: 344.84 Physical Form: Powder Appearance: White to off-white Melting Point: 141-149° Solubility: Soluble in chloroform, DMSO, DMF and alcohols MDL No.: MFCD00057220


Applications: Specific inhibitor of calcium-activated potassium channels

Formel: C₂₂H₁₇ClN₂
Schmelzpunkt: 141…149 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00057220
CAS-Nummer: 23593-75-1
EINECS: 245-764-8
Merck Index: 14,2417

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR