Aztreonam
Lieferant: Thermo Fisher Scientific
Synonyme:
2-[[(Z)-[1-(2-Amino-4-thiazolyl)-2-[[(2S,3S)-2-methyl-4-oxo-1-sulpho-3-azetidinyl]amino]-2-oxoethylidene]amino]oxy]-2-methylpropionic Acid, 2-((((Z)-1-(2-Aminothiazol-4-yl)-2-(((2S,3S)-2-methyl-4-oxo-1-sulphoazetidin-3-yl)amino)-2-oxoethylidene)amino)oxy)-2-methylpropanoic acid
CAS No.: 78110-38-0
Molecular Formula: C13H17N5O8S2
Formula Weight: 435.43
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Off-white
Melting Point: 227° dec.
Solubility: Soluble in DMF/water (1:1) at 50 mg/ml
MDL No.: MFCD00072145
Applications: Synthetic monocyclic beta-lactam antibiotic. Inhibits cell wall synthesis in Gram-negative bacteria
Formel:
C₁₃H₁₇N₅O₈S₂ Schmelzpunkt: 227 °C (dec.) |
MDL Number:
MFCD00072145 CAS-Nummer: 78110-38-0 EINECS: 278-839-9 Merck Index: 14,925 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...