Cefotaxim Natriumsalz

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Sodium (6R,7R)-3-(acetoxymethyl)-7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Cefotaxime sodium

J62690.03 J62690.06 J62690.14
J62690.03EA 120 EUR
J62690.03 J62690.06 J62690.14
Cefotaxim Natriumsalz
Cefotaxime sodium salt

CAS No.: 64485-93-4 Molecular Formula: C16H16N5NaO7S2 Formula Weight: 477.40 Physical Form: Powder Appearance: White to pale yellow Solubility: Freely soluble in water Beilstein Reference: 5711411 MDL No.: MFCD00079073

Formel: C₁₆H₁₇N₅O₇S₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00079073
CAS-Nummer: 64485-93-4
EINECS: 264-915-9
Merck Index: 14,1933

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR