Fast Blue BB base

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4'-Amino-2',5'-diethoxybenzanilide, N-(4-Amino-2,5-diethoxyphenyl)benzamide, Azoic Diazo No. 20, Fast blue BB, C.I. 37175, Azoic diazo component 20 (base)

J62456.14 J62456.22
J62456.14EA 62.8 EUR
J62456.14 J62456.22
Fast Blue BB base
Fast Blue BB base

CAS No.: 120-00-3 Molecular Formula: C17H20N2O3 Formula Weight: 300.35 Physical Form: Powder Appearance: Gray to pale brown Melting Point: 95-105° Beilstein Reference: 3416889 MDL No.: MFCD00009091


Applications: C.I. 37175

Formel: C₁₇H₂₀N₂O₃
Molekulargewicht: 300.36 g/mol
Schmelzpunkt: 95…105 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00009091
CAS-Nummer: 120-00-3
EINECS: 204-363-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR