Paromomycin sulphate
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulphate(1:x)
Inhibits initiation and elongation during protein synthesis.
CAS No.: 1263-89-4
Molecular Formula: C23H45N5O14.H2SO4
Formula Weight: 713.71
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in water
Beilstein Reference: 5715182
Optical Rotation: +51° (c=5 in water)
MDL No.: MFCD00079278
Applications: Inhibits initiation and elongation during protein synthesis
Formel:
C₂₃H₄₅N₅O₁₄·H₂SO₄ Molekulargewicht: 615.6 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00079278 CAS-Nummer: 1263-89-4 EINECS: 215-031-7 |