U0126 ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

J61246.MB J61246.MC
J61246.MBEA 172 EUR
J61246.MB J61246.MC
U0126 ≥99%
U0126

Selective inhibitor of mitogen-activated protein kinases MEK-1 and MEK-2.


  • Soluble in DMSO. Poorly soluble in ethanol and water

Formel: C₁₈H₁₆N₆S₂
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD01861186
CAS-Nummer: 109511-58-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR