Novobiocin sodium salt

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Sodium 4-((7-(((2R,3R,4S,5R)-4-(carbamoyloxy)-3-hydroxy-5-methoxy-6,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-4-hydroxy-8-methyl-2-oxo-2H-chromen-3-yl)carbamoyl)-2-(3-methylbut-2-en-1-yl)phenolate, Novobiocin sodium

J60928.03 J60928.06 J60928.09
J60928.03EA 49.5 EUR
J60928.03 J60928.06 J60928.09
Novobiocin sodium salt
Novobiocin sodium salt

Topoisomerase II inhibitor.


Novobiocin sodium salt is effective an antistaphylococcal agent used in the treatment of Methicillin-resistant Staphylococcus aureus (MRSA). It is also used to inhibit the activity of retroviral RNA-dependent DNA polymerase as well as inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase.


Applications: Topoisomerase II inhibitor

Formel: C₃₁H₃₅N₂NaO₁₁
Schmelzpunkt: 215 °C
MDL Number: MFCD00066541
CAS-Nummer: 1476-53-5
EINECS: 216-023-6
Merck Index: 14,6722

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR