Thymidine-5'-monophosphate disodium salt
Lieferant: Thermo Fisher Scientific
J60747.MC
J60747.MD
J60747.03
J60747.MCEA
46.9
EUR
J60747.MC
J60747.MD
J60747.03
Thymidine-5'-monophosphate disodium salt
Thymidine-5'-monophosphate disodium salt
CAS No.: 33430-62-5
Molecular Formula: C10H13N2Na2O8P
Formula Weight: 366.18
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in water
Beilstein Reference: 4086927
MDL No.: MFCD09039259
Formel:
C₁₀H₁₃N₂Na₂O₈P Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD09039259 CAS-Nummer: 33430-62-5 EINECS: 251-518-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...