Irinotecan hydrochloride
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(S)-4,11-Diethyl-4-hydroxy-3,14-dioxo-3,4,12,14-tetrahydro-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl [1,4'-bipiperidine]-1'-carboxylate hydrochloride
CAS No.: 100286-90-6
Molecular Formula: C33H38N4O6.HCl
Formula Weight: 623.14
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Yellow
Solubility: Soluble in DMSO or DMF at approximately 20mg/ml. Sparingly soluble in aqueous buffers.
MDL No.: MFCD01862255
Applications: Topoisomerase I inhibitor
Formel:
C₃₃H₃₈N₄O₆·HCl |
MDL Number:
MFCD01862255 CAS-Nummer: 100286-90-6 Merck Index: 14,5091 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...