Fmoc-D-Lys(Alloc)-OH 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Nepsilon-Allyloxycarbonyl-Nα-Fmoc-D-lysine, Fmoc-D-Lys(Aloc)-OH
H66861.03
H66861.06
H66861.03EA
53.4
EUR
H66861.03
H66861.06
Fmoc-D-Lys(Alloc)-OH 95%
Fmoc-D-Lys(Alloc)-OH
Purity: 95%
CAS No.: 214750-75-1
Molecular Formula: C25H28N2O6
Formula Weight: 452.51
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00798639
Formel:
C₂₅H₂₈N₂O₆ Molekulargewicht: 452.507 g/mol |
MDL Number:
MFCD00798639 CAS-Nummer: 214750-75-1 |