3-Chloro-N-fmoc-L-phenylalanine 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Fmoc-Phe(3-Cl)-OH
H66237.03
H66237.06
H66237.03EA
62.4
EUR
H66237.03
H66237.06
3-Chloro-N-fmoc-L-phenylalanine 95%
3-Chloro-N-fmoc-L-phenylalanine
Purity: 95%
CAS No.: 198560-44-0
Molecular Formula: C24H20ClNO4
Formula Weight: 421.88
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD00672548
Formel:
C₂₄H₂₀ClNO₄ Molekulargewicht: 421.877 g/mol |
MDL Number:
MFCD00672548 CAS-Nummer: 198560-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...