3-Brom-5-nitrobenzaldehyd 97+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H64787.03
H64787.06
H64787.03EA
80.8
EUR
H64787.03
H64787.06
3-Brom-5-nitrobenzaldehyd 97+%
3-Bromo-5-nitrobenzaldehyde
Purity: 97+%
CAS No.: 355134-13-3
Molecular Formula: C7H4BrNO3
Formula Weight: 230.02
Physical Form: Powder
Appearance: Pale brown
Melting Point: 101-106°
Beilstein Reference: 3253229
MDL No.: MFCD00100097
Formel:
C₇H₄BrNO₃ Schmelzpunkt: 101…106 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00100097 CAS-Nummer: 355134-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...