Methyl-5-brom-6-chlornicotinat 98%

Lieferant: Thermo Fisher Scientific
Achtung

H64754.06 H64754.14
H64754.06EA 102 EUR
H64754.06 H64754.14
Methyl-5-brom-6-chlornicotinat 98%
Methyl-5-brom-6-chlornicotinat

Purity: 98% CAS No.: 78686-77-8 Molecular Formula: C7H5BrClNO2 Formula Weight: 250.48 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 76-79° Beilstein Reference: 4403950 MDL No.: MFCD03844855

Formel: C₇H₅BrClNO₂
Schmelzpunkt: 76…79 °C
MDL Number: MFCD03844855
CAS-Nummer: 78686-77-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR