3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H64748.03
H64748.06
H64748.03EA
94.1
EUR
H64748.03
H64748.06
3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine 97%
3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine
Purity: 97%
CAS No.: 1031929-01-7
Molecular Formula: C6H2BrF4N
Formula Weight: 243.99
Flash Point: 30°(86°F)
Physical Form: Liquid
Density: 1.774
Refractive Index: 1.4600
UN No.: UN1992
MDL No.: MFCD08061304
Formel:
C₆H₂BrF₄N Dichte: 1.774 Flammpunkt: 30 °C (86 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08061304 CAS-Nummer: 1031929-01-7 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...