Methyl-3-fluor-4-nitrobenzoat 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl 3-fluoro-4-nitrobenzoate, Methyl 3-fluoro-4-nitrobenzenecarboxylate, 3-Fluorobenzoic acid methyl ester
H64688.06
H64688.14
H64688.06EA
67.5
EUR
H64688.06
H64688.14
Methyl-3-fluor-4-nitrobenzoat 97%
Methyl-3-fluor-4-nitrobenzoat
Purity: 97%
CAS No.: 185629-31-6
Molecular Formula: C8H6FNO4
Formula Weight: 199.14
Physical Form: Solid
Beilstein Reference: 10657755
MDL No.: MFCD08444027
Formel:
C₈H₆FNO₄ Molekulargewicht: 199.14 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08444027 CAS-Nummer: 185629-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...