4,4'-Dibromotriphenylamine 98%

Lieferant: Thermo Fisher Scientific
Achtung

H64666.03 H64666.06
H64666.03EA 67 EUR
H64666.03 H64666.06
4,4'-Dibromotriphenylamine 98%
4,4'-Dibromotriphenylamine

Purity: 98% CAS No.: 81090-53-1 Molecular Formula: C18H13Br2N Formula Weight: 403.12 Flash Point: 110°(230°F) Physical Form: Solid Appearance: Off-white Melting Point: 85° Beilstein Reference: 7094354 MDL No.: MFCD00060104

Formel: C₁₈H₁₃Br₂N
Schmelzpunkt: 85 °C
Flammpunkt: >110 °C (230 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00060104
CAS-Nummer: 81090-53-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR