2-Bromo-6-hydrazinylpyridine 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Bromo-6-hydrazinopyridine

H64664.06 H64664.14
H64664.06EA 111 EUR
H64664.06 H64664.14
2-Bromo-6-hydrazinylpyridine 98%
2-Bromo-6-hydrazinylpyridine

Purity: 98% CAS No.: 26944-71-8 Molecular Formula: C5H6BrN3 Formula Weight: 188.03 Physical Form: Solid Appearance: Pale brown Melting Point: 117-118° Beilstein Reference: 386966 UN No.: UN2811 MDL No.: MFCD09743754

Formel: C₅H₆BrN₃
Schmelzpunkt: 117…118 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09743754
CAS-Nummer: 26944-71-8
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR