L-Lysine ethyl ester diisocyanate 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
(S)-Ethyl 2,6-diisocyanatohexanoate
H64384.06
H64384.14
H64384.06EA
152
EUR
H64384.06
H64384.14
L-Lysine ethyl ester diisocyanate 97%
L-Lysine ethyl ester diisocyanate
Purity: 97%
CAS No.: 45172-15-4
Molecular Formula: C10H14N2O4
Formula Weight: 226.23
Storage Temperature: 2°C to 8°C
Physical Form: Viscous liquid
Refractive Index: 1.4572
Sensitivity: Moisture Sensitive
Appearance: Yellow to brown
Optical Rotation: -35° (c=1 in dioxane)
MDL No.: MFCD08276480
Formel:
C₁₀H₁₄N₂O₄ |
MDL Number:
MFCD08276480 CAS-Nummer: 45172-15-4 UN-Nummer: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...